ENA370563133 Enamine

2-methyl-1,3-benzoxazole-5-sulfonyl chloride

  • MDL number MFCD11650658
  • Cas Number : 1094671-83-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 181...183
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105170

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105170
Availability details for
Quantity
  • Availability
  • Add to Cart