ENA370563134 Enamine

[2-(furan-2-ylmethoxy)-4-methylphenyl]methanamine

  • MDL number MFCD16731715
  • Cas Number : 1248059-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105171

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105171
Availability details for
Quantity
  • Availability
  • Add to Cart