ENA370563141 Enamine

4-tert-butyl-2-(chloromethyl)-1,3-thiazole

  • MDL number MFCD13189373
  • Cas Number : 170881-59-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105179

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105179
Availability details for
Quantity
  • Availability
  • Add to Cart