ENA370563157 Enamine

N-(2-ethylcyclohexyl)-2-fluoropyridine-4-carboxamide

  • MDL number MFCD13338488
  • Cas Number : 1308368-28-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69...71
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103320

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103320
Availability details for
Quantity
  • Availability
  • Add to Cart