ENA370563169 Enamine

4-(2-chlorophenyl)-1,3-oxazol-2-amine

  • MDL number MFCD16667974
  • Cas Number : 1248965-63-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 137...139
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105283

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-105283
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart