ENA370563173 Enamine

2-fluoro-N-(1-methylcyclopentyl)pyridine-4-carboxamide

  • MDL number MFCD14643627
  • Cas Number : 1249421-71-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-103327

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-103327
Availability details for
Quantity
  • Availability
  • Add to Cart