ENA370563186 Enamine

6-(4-chlorophenoxy)pyridine-2-carboxylic acid

  • MDL number MFCD17318826
  • Cas Number : 51362-41-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115...117
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103449

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103449
Availability details for
Quantity
  • Availability
  • Add to Cart