ENA370563187 Enamine

2-fluoro-5-[(4-methoxypiperidin-1-yl)carbonylamino]benzoic acid

  • MDL number MFCD22369900
  • Cas Number : 1307190-62-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105306

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105306
Availability details for
Quantity
  • Availability
  • Add to Cart