ENA370563196 Enamine

1H-imidazole-5-carbohydrazide

  • MDL number MFCD09839062
  • Cas Number : 18329-78-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105316

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105316
Availability details for
Quantity
  • Availability
  • Add to Cart