ENA370563202 Enamine

3-chloro-4-methoxy-N-methylaniline

  • MDL number MFCD11151134
  • Cas Number : 90234-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105324

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105324
Availability details for
Quantity
  • Availability
  • Add to Cart