ENA370563208 Enamine

5-[(4-methoxybenzene)amido]thiophene-2-carboxylic acid

  • MDL number MFCD12579962
  • Cas Number : 1152992-43-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 198...200
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105331

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105331
Availability details for
Quantity
  • Availability
  • Add to Cart