ENA370563223 Enamine

2-[2-bromo-6-ethoxy-4-(hydroxymethyl)phenoxy]acetamide

  • MDL number MFCD05862194
  • Cas Number : 838904-26-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105381

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105381
Availability details for
Quantity
  • Availability
  • Add to Cart