ENA370563238 Enamine

methyl 2-[(3-nitropyridin-2-yl)amino]propanoate

  • MDL number MFCD12189084
  • Cas Number : 146294-95-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105405

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105405
Availability details for
Quantity
  • Availability
  • Add to Cart