ENA370563239 Enamine

methyl 3-[(3-nitropyridin-2-yl)amino]propanoate

  • MDL number MFCD12797437
  • Cas Number : 1183435-02-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-105410

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105410
Availability details for
Quantity
  • Availability
  • Add to Cart