ENA370563241 Enamine

1H-imidazo[4,5-b]pyridin-2-amine

  • MDL number MFCD03792824
  • Cas Number : 132898-03-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 268...270
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105412

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105412
Availability details for
Quantity
  • Availability
  • Add to Cart