ENA370563243 Enamine

1-bromocyclopentane-1-carboxylic acid

  • MDL number MFCD19301875
  • Cas Number : 56447-24-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66...68
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101126

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101126
Availability details for
Quantity
  • Availability
  • Add to Cart