ENA370563244 Enamine

2,6-dichloro-3-methanesulfonylbenzoic acid

  • MDL number MFCD11179846
  • Cas Number : 1094404-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105418

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105418
Availability details for
Quantity
  • Availability
  • Add to Cart