ENA370563252 Enamine

2-(2-amino-2-phenylethoxy)-1,3-dimethylbenzene

  • MDL number MFCD09731599
  • Cas Number : 639011-03-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105429

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105429
Availability details for
Quantity
  • Availability
  • Add to Cart