ENA370563255 Enamine

2,2-dimethyl-1-{2-methyl-octahydropyrrolo[2,3-c]pyrrol-1-yl}propan-1-one

  • MDL number MFCD12082303
  • Cas Number : 1218273-10-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105439

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105439
Availability details for
Quantity
  • Availability
  • Add to Cart