ENA370563258 Enamine

1,3,3-trimethylpiperazin-2-one hydrochloride

  • MDL number MFCD15072084
  • Cas Number : 1207877-86-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 209...211
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105468

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-105468
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart