ENA370563259 Enamine

[4-methyl-2-(propan-2-yloxy)phenyl]methanamine

  • MDL number MFCD16704892
  • Cas Number : 1247490-96-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105473

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105473
Availability details for
Quantity
  • Availability
  • Add to Cart