ENA370563260 Enamine

4-(chloromethyl)-1-methyl-5-(trifluoromethyl)-1H-pyrazole

  • MDL number MFCD21774655
  • Cas Number : 1384429-36-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105474

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105474
Availability details for
Quantity
  • Availability
  • Add to Cart