ENA370563272 Enamine

1H,2H,3H,4H,5H-pyrido[3,4-b][1,4]diazepin-2-one

  • MDL number MFCD08234946
  • Cas Number : 885271-88-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105503

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105503
Availability details for
Quantity
  • Availability
  • Add to Cart