ENA370563273 Enamine

2-methyl-1-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}pyrrolidine

  • MDL number MFCD18781106
  • Cas Number : 1486485-52-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103792

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-103792
Availability details for
Quantity
  • Availability
  • Add to Cart