ENA370563274 Enamine

4-chloro-2,3-dihydro-1H-indole hydrochloride

  • MDL number MFCD09026825
  • Cas Number : 1251023-48-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105505

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-105505
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart