ENA370563282 Enamine

methyl 3-amino-2,2-dimethylpropanoate hydrochloride

  • MDL number MFCD10758092
  • Cas Number : 177269-37-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158...160
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105537

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-105537
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart