ENA370563286 Enamine

2-amino-3,3-dimethylcyclopent-1-ene-1-carbonitrile

  • MDL number MFCD19206705
  • Cas Number : 3597-66-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105541

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105541
Availability details for
Quantity
  • Availability
  • Add to Cart