ENA370563292 Enamine

1-methyl-4-{2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy]ethyl}piperazine

  • MDL number MFCD09746225
  • Cas Number : 910462-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103822

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-103822
Availability details for
Quantity
  • Availability
  • Add to Cart