ENA370563296 Enamine

4-bromo-2-(methylsulfanyl)benzaldehyde

  • MDL number MFCD18887984
  • Cas Number : 918350-11-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88-89
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-105556

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105556
Availability details for
Quantity
  • Availability
  • Add to Cart