ENA370563299 Enamine

5-(piperidine-1-sulfonyl)-1,2-dihydropyridin-2-one

  • MDL number MFCD12096498
  • Cas Number : 627839-96-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105559

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105559
Availability details for
Quantity
  • Availability
  • Add to Cart