ENA370563300 Enamine

3-methyl-1-(2,2,2-trifluoroethyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD07357370
  • Cas Number : 113100-55-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105560

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105560
Availability details for
Quantity
  • Availability
  • Add to Cart