ENA370563303 Enamine

1,4-difluoro-2-isocyanato-3-methoxybenzene

  • MDL number MFCD18395264
  • Cas Number : 1261616-22-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105565

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105565
Availability details for
Quantity
  • Availability
  • Add to Cart