ENA370563304 Enamine

3-(3-chlorophenyl)-2,2-dimethylpropan-1-amine

  • MDL number MFCD16855896
  • Cas Number : 1267940-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105566

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105566
Availability details for
Quantity
  • Availability
  • Add to Cart