ENA370563311 Enamine

methyl 4-(bromomethyl)-2-fluorobenzoate

  • MDL number MFCD11976782
  • Cas Number : 85070-57-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 74...76
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105585

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-105585
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart