ENA370563312 Enamine

5-bromo-1-(3-chlorophenyl)-3-methyl-1H-pyrazolo[3,4-b]pyridine

  • MDL number MFCD11127771
  • Cas Number : 1019125-50-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105586

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105586
Availability details for
Quantity
  • Availability
  • Add to Cart