ENA370563317 Enamine

3-(2-methylpropanamido)naphthalene-2-carboxylic acid

  • MDL number MFCD09044674
  • Cas Number : 926234-29-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-105592

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105592
Availability details for
Quantity
  • Availability
  • Add to Cart