ENA370563322 Enamine

ethyl 2-azabicyclo[4.1.0]heptane-1-carboxylate

  • MDL number MFCD14581330
  • Cas Number : 1250993-25-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105630

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105630
Availability details for
Quantity
  • Availability
  • Add to Cart