ENA370563330 Enamine

[1-(2-methoxyethyl)pyrrolidin-2-yl]methanol

  • MDL number MFCD16163488
  • Cas Number : 1249698-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105649

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105649
Availability details for
Quantity
  • Availability
  • Add to Cart