ENA370563332 Enamine

2-(4-fluoro-2-methylphenyl)ethan-1-amine

  • MDL number MFCD08449245
  • Cas Number : 910400-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103956

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-103956
Availability details for
Quantity
  • Availability
  • Add to Cart