ENA370563334 Enamine

2-(piperazin-1-yl)-1-(pyrrolidin-1-yl)propan-1-one

  • MDL number MFCD10011187
  • Cas Number : 89574-40-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-104016

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104016
Availability details for
Quantity
  • Availability
  • Add to Cart