ENA370563338 Enamine

3-propylpiperidine-2,6-dione

  • MDL number MFCD02262200
  • Cas Number : 1341764-58-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76...78
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105657

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105657
Availability details for
Quantity
  • Availability
  • Add to Cart