ENA370563340 Enamine

4-[4-(difluoromethoxy)phenyl]-3-methyl-1H-pyrazol-5-amine

  • MDL number MFCD11208950
  • Cas Number : 1187766-74-0
  • Molecular Weight : 239.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-104038

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104038
Availability details for
Quantity
  • Availability
  • Add to Cart