ENA370563343 Enamine

3-chloro-4-sulfanylbenzonitrile

  • MDL number MFCD17290940
  • Cas Number : 38234-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-105667

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105667
Availability details for
Quantity
  • Availability
  • Add to Cart