ENA370563361 Enamine

1-ethylcyclopentane-1-carboxylic acid

  • MDL number MFCD16860495
  • Cas Number : 17206-19-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105710

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105710
Availability details for
Quantity
  • Availability
  • Add to Cart