ENA370563363 Enamine

3-(3-aminopropoxy)oxolane

  • MDL number MFCD18364653
  • Cas Number : 1339095-68-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105713

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105713
Availability details for
Quantity
  • Availability
  • Add to Cart