ENA370563372 Enamine

3-amino-N-methyl-4-sulfanylbenzene-1-sulfonamide

  • MDL number MFCD12173051
  • Cas Number : 1154232-59-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105723

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105723
Availability details for
Quantity
  • Availability
  • Add to Cart