ENA370563376 Enamine

5-[(3,4-dichlorophenyl)methyl]-2H-1,2,3,4-tetrazole

  • MDL number MFCD19352653
  • Cas Number : 1039864-56-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-105733

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105733
Availability details for
Quantity
  • Availability
  • Add to Cart