ENA370563380 Enamine

2-amino-4-methyl-5-pentylthiophene-3-carboxamide

  • MDL number MFCD11198349
  • Cas Number : 68746-23-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105738

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105738
Availability details for
Quantity
  • Availability
  • Add to Cart