ENA370563386 Enamine

2-(difluoromethoxy)-5-methylbenzaldehyde

  • MDL number MFCD18331348
  • Cas Number : 147249-30-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105747

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105747
Availability details for
Quantity
  • Availability
  • Add to Cart