ENA370563398 Enamine

3-amino-1-(2-methoxyphenyl)pyrrolidin-2-one

  • MDL number MFCD14689505
  • Cas Number : 1247859-12-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105820

Safety Information

msds https://www.enaminestore.com/catalog/EN300-105820
Availability details for
Quantity
  • Availability
  • Add to Cart